2-Fluoro-4-nitrotoluene ≥97.0%

Supplier: TCI America

F0292-25G F0292-5G
TCF0292-25GEA 120.52 CAD
TCF0292-25G TCF0292-5G
2-Fluoro-4-nitrotoluene ≥97.0%
2-Fluoro-4-nitrotoluene

CAS Number: 1427-07-2
MDL Number: MFCD00007199
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 73
Freezing point (°C): 33

Formula: C₇H₆FNO₂
Flash Pt: 73 °C
MDL Number: MFCD00007199
CAS Number: 1427-07-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba7832d8c4de254c0e411aa6746b25eff8eb760a&vid=f61003596ec42878b573989e17893a3aae5ae609&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR