2-Fluoro-L-phenylalanine ≥96.0%

Supplier: TCI America

Synonyms: H-Phe(2-F)-OH

F0273-1G F0273-100MG
TCF0273-1GEA 235.42 CAD
TCF0273-1G TCF0273-100MG
2-Fluoro-L-phenylalanine ≥96.0%
2-Fluoro-L-phenylalanine

CAS Number: 19883-78-4
MDL Number: MFCD00066448
Molecular Formula: C9H10FNO2
Molecular Weight: 183.18
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 217
Specific rotation [a]20/D: -15 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-3-(2-fluorophenyl)propionic Acid, H-Phe(2-F)-OH

Formula: C₉H₁₀FNO₂
Melting Pt: 217 °C
MDL Number: MFCD00066448
CAS Number: 19883-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=170b731d94223eaa6cb34fc96e7d08aa535eb24d&vid=170b731d94223eaa6cb34fc96e7d08aa535eb24d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR