2-Fluorobenzyl alcohol ≥97.0%

Supplier: TCI America

Synonyms: (2-Fluorophenyl)methanol, 2-Fluorobenzyl Alcohol

F0259-25G F0259-5G
TCF0259-25GEA 139.04 CAD
TCF0259-25G TCF0259-5G
2-Fluorobenzyl alcohol ≥97.0%
2-Fluorobenzyl alcohol

CAS Number: 446-51-5
MDL Number: MFCD00004601
Molecular Formula: C7H7FO
Molecular Weight: 126.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 90
Specific Gravity (20/20): 1.18

Formula: C₇H₇FO
Boiling Pt: 96 °C
Density: 1.18 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00004601
CAS Number: 446-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a46bbb8274b06ab28652426ff073b3e72c6b5ac&vid=3a46bbb8274b06ab28652426ff073b3e72c6b5ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR