Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
CAS Number: 82911-69-1
MDL Number: MFCD00010733
Molecular Formula: C19H15NO5
Molecular Weight: 337.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 153
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: 9-Fluorenylmethyl N-Succinimidyl Carbonate, N-(Fmoc-oxy)succinimide, Carbonic Acid 9-Fluorenylmethyl N-Succinimidyl Ester, Fmoc-OSu
Formula:
C₁₉H₁₅NO₅ MW: 337.3 g/mol Melting Pt: 153 °C |
MDL Number:
MFCD00010733 CAS Number: 82911-69-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...