4-(2-Furyl)-3-buten-2-one ≥98.0%

Supplier: TCI America

F0235-25G F0235-100G
TCF0235-025GEA 161.16 CAD
TCF0235-025G TCF0235-100G
4-(2-Furyl)-3-buten-2-one ≥98.0%
4-(2-Furyl)-3-buten-2-one

CAS Number: 623-15-4
MDL Number: MFCD00039566
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystalline Powder
Flash Point (°C): 110
Freezing point (°C): 37
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Furfuralacetone

Formula: C₈H₈O₂
Flash Pt: 110 °C
MDL Number: MFCD00039566
CAS Number: 623-15-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da07acfda5af340aca9696c23ef1f181ce88bdfc&vid=da07acfda5af340aca9696c23ef1f181ce88bdfc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR