Fluorescamine
Supplier: TCI America
Synonyms:
Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
F0192-1G
F0192-100MG
TCF0192-1GEA
723.96
CAD
TCF0192-1G
TCF0192-100MG
Fluorescamine
Fluorescamine
[for HPLC Labeling]
CAS Number: 38183-12-9
MDL Number: MFCD00005928
Molecular Formula: C17H10O4
Molecular Weight: 278.26
Form: Crystal
Color: White
Melting point (°C): 155
Other Product Names / Synonyms: 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
Formula:
C₁₇H₁₀O₄ MW: 278.26 g/mol Melting Pt: 155 °C |
MDL Number:
MFCD00005928 CAS Number: 38183-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...