Tartrazine ≥98.0% (by HPLC)
Supplier: TCI America
F0088-500G
F0088-25G
TCF0088-500GEA
371.3
CAD
TCF0088-500G
TCF0088-025G
Tartrazine ≥98.0% (by HPLC)
Tartrazine
CAS Number: 1934-21-0
MDL Number: MFCD00148908
Molecular Formula: C16H12N4O9S2
Molecular Weight: 534.36
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Lambda max.: 428 nm (AcONH4 sol.)
Other Product Names / Synonyms: Filter Yellow, Tartrazine
Formula:
C₁₆H₁₂N₄O₉S₂ |
MDL Number:
MFCD00148908 CAS Number: 1934-21-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...