2-Furoyltrifluoroacetone

Supplier: TCI America

Synonyms: 4,4,4-Trifluoro-1-(furan-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione

F0083-25G F0083-5G
TCF0083-025GEA 226.41 CAD
TCF0083-025G TCF0083-005G
2-Furoyltrifluoroacetone
2-Furoyltrifluoroacetone

CAS Number: 326-90-9
MDL Number: MFCD00020935
Molecular Formula: C8H5F3O3
Molecular Weight: 206.12
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 84


Other Product Names / Synonyms: 4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 84 °C
MDL Number: MFCD00020935
CAS Number: 326-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aeb2085adf2ff1110cb85aa6471bc37c29a10d24&vid=aeb2085adf2ff1110cb85aa6471bc37c29a10d24&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR