Furil ≥98.0%

Supplier: TCI America

F0078-5G F0078-25G
TCF0078-005GEA 229.1 CAD
TCF0078-005G TCF0078-025G
Furil ≥98.0%
Furil

CAS Number: 492-94-4
MDL Number: MFCD00003241
Molecular Formula: C10H6O4
Molecular Weight: 190.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 164

Formula: C₁₀H₆O₄
Melting Pt: 164 °C
MDL Number: MFCD00003241
CAS Number: 492-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=929968a66687109c15c3094a77d950d980442119&vid=929968a66687109c15c3094a77d950d980442119&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR