Sodium hydrogen fumarate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Monosodium Fumarate, Fumaric Acid Monosodium Salt
F0071-25G
F0071-500G
TCF0071-025GEA
80.58
CAD
TCF0071-025G
TCF0071-500G
Sodium hydrogen fumarate ≥98.0% (by titrimetric analysis)
Sodium hydrogen fumarate
CAS Number: 5873-57-4
MDL Number: MFCD00039097
Molecular Formula: C4H4O4
Molecular Weight: 138.05
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Monosodium Fumarate, Fumaric Acid Monosodium Salt
Formula:
C₄H₄O₄ MW: 138.05 g/mol |
MDL Number:
MFCD00039097 CAS Number: 5873-57-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...