L-(-)-Fucose ≥97.0% (by HPLC)
Supplier: TCI America
F0065-1G
F0065-10G
TCF0065-001GEA
77.12
CAD
TCF0065-001G
TCF0065-010G
L-(-)-Fucose ≥97.0% (by HPLC)
L(-)-Fucose
CAS Number: 2438-80-4
MDL Number: MFCD00135607
Molecular Formula: C6H12O5
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 140
Specific rotation [a]20/D: -75 deg (C=1, H2O)
Product Categories: Bioscience, Fucose, Deoxysugars
Other Product Names / Synonyms: 6-Deoxy-L-galactose, L-(-)-Rhodeose
Formula:
C₆H₁₂O₅ Melting Pt: 140 °C |
MDL Number:
MFCD00069812 CAS Number: 2438-80-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...