Ferroin (1,10-phenanthrolineferrous sulfate complex) in solution acc. JIS K8001

Supplier: TCI America

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate, Tris(1,10-phenanthroline)iron (II) sulfate, Tris(1,10-phenanthroline-N1,N10)iron sulfate

F0005-25G F0005-500G
TCF0005-025GEA 44.24 CAD
TCF0005-025G TCF0005-500G
Ferroin (1,10-phenanthrolineferrous sulfate complex) in solution acc. JIS K8001
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)

CAS Number: 14634-91-4 MDL Number: MFCD00036428 Form: Liquid Color: Yellow Red


Form: Liquid
Color: Yellow Red

Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
MDL Number: MFCD00036428
CAS Number: 14634-91-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR