N-(Ethoxycarbonyl)nortropinone ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, 8-(Ethoxycarbonyl)-8-azabicyclo[3.2.1]octan-3-one, N-Carbethoxy-4-tropinone

E0834-5G E0834-25G
TCE0834-5GEA 248.06 CAD
TCE0834-5G TCE0834-25G
N-(Ethoxycarbonyl)nortropinone ≥98.0%
N-(Ethoxycarbonyl)nortropinone

CAS Number: 32499-64-2
MDL Number: MFCD00078171
Molecular Formula: C10H15NO3
Molecular Weight: 197.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Specific Gravity (20/20): 1.15


Product Categories: Bioscience, Tropane Alkaloids
Other Product Names / Synonyms: 8-(Ethoxycarbonyl)-8-azabicyclo[3.2.1]octan-3-one

Formula: C₁₀H₁₅NO₃
MW: 197.23 g/mol
Boiling Pt: 153 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00078171
CAS Number: 32499-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6d64d5c33a44782a835c6981d82517653fd048e&vid=a6d64d5c33a44782a835c6981d82517653fd048e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR