1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥98.0%

Supplier: TCI America

Synonyms: 1-(4-Ethylphenyl)-2-(4-methoxyphenyl)acetylene, 4-[(4-Ethylphenyl)ethynyl]anisole, 1-Ethyl-4-((4-methoxyphenyl)ethynyl)benzene

E0806-5G E0806-1G
TCE0806-5GEA 200.66 CAD
TCE0806-5G TCE0806-1G
1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥98.0%
1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene

CAS Number: 63221-88-5
MDL Number: MFCD00807428
Molecular Formula: C17H16O
Molecular Weight: 236.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 153
Melting point (°C): 74


Other Product Names / Synonyms: 4-[(4-Ethylphenyl)ethynyl]anisole, 1-(4-Ethylphenyl)-2-(4-methoxyphenyl)acetylene

Formula: C₁₇H₁₆O
Boiling Pt: 153 °C
Melting Pt: 74 °C
MDL Number: MFCD00807428
CAS Number: 63221-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a51097d9dc793482d3c68a893032e07b84388efa&vid=a51097d9dc793482d3c68a893032e07b84388efa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR