EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥95.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI

E0805-25G E0805-5G
TCE0805-25GEA 175.38 CAD
TCE0805-25G TCE0805-5G
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥95.0% (by titrimetric analysis) for biochemical research
EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)

[for Biochemical Research]
CAS Number: 67-42-5
MDL Number: MFCD00004291
Molecular Formula: C14H24N2O10
Molecular Weight: 380.35
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 241


Product Categories: Bioscience, Protease Inhibitors (for Biochemical Research), Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Glycoletherdiaminetetraacetic Acid, 1,2-Bis[2-[bis(carboxymethyl)amino]ethoxy]ethane, EGTA, GEDTA

Formula: C₁₄H₂₄N₂O₁₀
MW: 380.35 g/mol
Melting Pt: 241 °C
MDL Number: MFCD00004291
CAS Number: 67-42-5
Merck Index: 13,03560
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4098991b1e7cc7671c8b0676475a949f3bb8595e&vid=2a08616b911a035d44d2262afa4c579479e754e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR