Ethyl-3-hydroxyhexanoate ≥98.0%

Supplier: TCI America

E0788-100G E0788-25G
TCE0788-100GEA 289.14 CAD
TCE0788-100G TCE0788-25G
Ethyl-3-hydroxyhexanoate ≥98.0%
Ethyl 3-hydroxyhexanoate

CAS Number: 2305-25-1
MDL Number: MFCD00036604
Molecular Formula: C8H16O3
Molecular Weight: 160.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 94
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: 3-Hydroxyhexanoic Acid Ethyl Ester

Formula: C₈H₁₆O₃
Boiling Pt: 92 °C
Density: 0.98 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00036604
CAS Number: 2305-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9bcac6dba02f5985013211de18d491da362e9ad&vid=a9bcac6dba02f5985013211de18d491da362e9ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR