Ethyl-2-Amino-4-phenylthiazole-5-carboxylate ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

E0769-1G E0769-5G
TCE0769-1GEA 127.98 CAD
TCE0769-1G TCE0769-5G
Ethyl-2-Amino-4-phenylthiazole-5-carboxylate ≥97.0% (by HPLC, titration analysis)
Ethyl-2-Amino-4-phenylthiazole-5-carboxylate

CAS Number: 64399-23-1
MDL Number: MFCD00022452
Molecular Formula: C12H12N2O2S
Molecular Weight: 248.30
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 169


Other Product Names / Synonyms: 2-Amino-4-phenylthiazole-5-carboxylic Acid Ethyl Ester

Formula: C₁₂H₁₂N₂O₂S
Melting Pt: 169 °C
MDL Number: MFCD00022452
CAS Number: 64399-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8deec72fd495f0f940414fef4d467530b3417759&vid=8deec72fd495f0f940414fef4d467530b3417759&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR