1-Propyl-4-ethynylbenzene ≥96.0%

Supplier: TCI America

E0750-5G E0750-25G
TCE0750-5GEA 79 CAD
TCE0750-5G TCE0750-25G
1-Propyl-4-ethynylbenzene ≥96.0%
1-Propyl-4-ethynylbenzene

CAS Number: 62452-73-7
MDL Number: MFCD00173886
Molecular Formula: C11H12
Molecular Weight: 144.22
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.91


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Propylphenylacetylene

Formula: C₁₁H₁₂
Density: 0.91 (20 °C)
MDL Number: MFCD00173886
CAS Number: 62452-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c368f23882a3f41fed61ce2d213d702e988b02d0&vid=c368f23882a3f41fed61ce2d213d702e988b02d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR