4-Ethylphenylacetylene ≥97.0%

Supplier: TCI America

Synonyms: (4-Ethylphenyl)acetylene, 1-Ethyl-4-ethynylbenzene, 1-Ethynyl-4-ethylbenzene, (4-Ethylphenyl)ethyne

E0749-5G E0749-25G
TCE0749-5GEA 211.72 CAD
TCE0749-5G TCE0749-25G
4-Ethylphenylacetylene ≥97.0%
4-Ethylphenylacetylene

CAS Number: 40307-11-7
MDL Number: MFCD00173887
Molecular Formula: C10H10
Molecular Weight: 130.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Flash Point (°C): 71
Specific Gravity (20/20): 0.92


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Ethylphenylacetylene

Formula: C₁₀H₁₀
MW: 130.19 g/mol
Boiling Pt: 192 °C
Density: 0.92 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00173887
CAS Number: 40307-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60da93f749d8b88a76d0695132ef973dcdca3739&vid=60da93f749d8b88a76d0695132ef973dcdca3739&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR