Ethyl-4-hydroxy-3-methoxycinnamate ≥97.0%

Supplier: TCI America

E0739-25G E0739-5G
TCE0739-25GEA 170.36 CAD
TCE0739-25G TCE0739-5G
Ethyl-4-hydroxy-3-methoxycinnamate ≥97.0%
Ethyl-4-hydroxy-3-methoxycinnamate

CAS Number: 4046-02-0
MDL Number: MFCD00009190
Molecular Formula: C12H14O4
Molecular Weight: 222.24
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 57


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 4-Hydroxy-3-methoxycinnamic Acid Ethyl Ester, Ethyl Ferulate, Ferulic Acid Ethyl Ester

Formula: C₁₂H₁₄O₄
Boiling Pt: 185 °C
Melting Pt: 57 °C
MDL Number: MFCD00009190
CAS Number: 4046-02-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d442e43922239becc8a378bc71d3aef722858aa&vid=9d442e43922239becc8a378bc71d3aef722858aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR