Ethyl thiooxamate ≥98.0%

Supplier: TCI America

Synonyms: 2-Thiooxamic acid ethyl ester, Ethyl amino(thioxo)acetate, Ethyl 2-amino-2-thioxoacetate

E0735-1G E0735-5G
TCE0735-1GEA 47.4 CAD
TCE0735-1G TCE0735-5G
Ethyl thiooxamate ≥98.0%
Ethyl thiooxamate

CAS Number: 16982-21-1
MDL Number: MFCD00074903
Molecular Formula: C4H7NO2S
Molecular Weight: 133.17
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 64
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Thiooxamic Acid Ethyl Ester, Ethyl Aminothioxoacetate, Aminothioxoacetic Acid Ethyl Ester

Formula: C₄H₇NO₂S
MW: 133.17
Melting Pt: 64 °C
MDL Number: MFCD00074903
CAS Number: 16982-21-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=382ad7a3a4aa707234ef40b3f678e27627f9acd4&vid=382ad7a3a4aa707234ef40b3f678e27627f9acd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR