4-Ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

E0725-5G E0725-1G
TCE0725-5GEA 94.8 CAD
TCE0725-5G TCE0725-1G
4-Ethoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Ethoxyphenylboronic acid

CAS Number: 22237-13-4
MDL Number: MFCD00674028
Molecular Formula: C8H11BO3
Molecular Weight: 165.98
Form: Crystal
Color: White
Melting point (°C): 168


Other Product Names / Synonyms: 4-Ethoxybenzeneboronic Acid

Formula: C₈H₁₁BO₃
Melting Pt: 168 °C
MDL Number: MFCD00674028
CAS Number: 22237-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03303d5ecc2d8c4b3c6368fdefd8356e3992f5f4&vid=03303d5ecc2d8c4b3c6368fdefd8356e3992f5f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR