3-Ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

E0724-1G E0724-5G
TCE0724-1GEA 34.76 CAD
TCE0724-1G TCE0724-5G
3-Ethoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Ethoxyphenylboronic acid

CAS Number: 90555-66-1
MDL Number: MFCD00274219
Molecular Formula: C8H11BO3
Molecular Weight: 165.98
Form: Crystal
Color: White
Melting point (°C): 153


Other Product Names / Synonyms: 3-Ethoxybenzeneboronic Acid

Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00274219
CAS Number: 90555-66-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93fda71bf6617edc3ad4239d6b131d0c3350b556&vid=93fda71bf6617edc3ad4239d6b131d0c3350b556&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR