4-Ethylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
E0720-5G
E0720-25G
TCE0720-5GEA
47.4
CAD
TCE0720-5G
TCE0720-25G
4-Ethylphenylboronic acid (contains varying amounts of Anhydride)
4-Ethylphenylboronic acid
CAS Number: 63139-21-9
MDL Number: MFCD00859377
Molecular Formula: C8H11BO2
Molecular Weight: 149.98
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 141
Other Product Names / Synonyms: 4-Ethylbenzeneboronic Acid
Formula:
C₈H₁₁BO₂ Melting Pt: 141 °C |
MDL Number:
MFCD00859377 CAS Number: 63139-21-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...