4-Ethylbenzyl alcohol ≥98.0%
Supplier: TCI America
Synonyms:
(4-Ethylphenyl)methanol
E0704-25G
E0704-5G
TCE0704-25GEA
522.35
CAD
TCE0704-25G
TCE0704-5G
4-Ethylbenzyl alcohol ≥98.0%
4-Ethylbenzyl alcohol
CAS Number: 768-59-2
MDL Number: MFCD00004666
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 104
Specific Gravity (20/20): 1.00
Formula:
C₉H₁₂O Boiling Pt: 117 °C Density: 1.00 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00004666 CAS Number: 768-59-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...