Ethyl-5-chloro-1H-indole-2-carboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl 5-chloro-1H-indole-2-carboxylate, Ethyl 5-chloro-2-indolecarboxylate
E0696-5G
E0696-25G
TCE0696-5GEA
142.2
CAD
TCE0696-5G
TCE0696-25G
Ethyl-5-chloro-1H-indole-2-carboxylate ≥98.0%
Ethyl-5-chloro-1H-indole-2-carboxylate
CAS Number: 4792-67-0
MDL Number: MFCD00005610
Molecular Formula: C11H10ClNO2
Molecular Weight: 223.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 170
Other Product Names / Synonyms: 5-Chloroindole-2-carboxylic Acid Ethyl Ester
Formula:
C₁₁H₁₀ClNO₂ MW: 223.66 g/mol Melting Pt: 170 °C |
MDL Number:
MFCD00005610 CAS Number: 4792-67-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...