1-Ethyl-3-methylimidazolium ethylsulfate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

E0650-25G E0650-5G
TCE0650-25GEA 252.8 CAD
TCE0650-25G TCE0650-5G
1-Ethyl-3-methylimidazolium ethylsulfate ≥98.0% (by total nitrogen basis)
1-Ethyl-3-methylimidazolium ethyl sulphate

CAS Number: 342573-75-5
Molecular Formula: C8H16N2O4S
Molecular Weight: 236.29
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Flash Point (°C): 162
Specific Gravity (20/20): 1.24

Formula: C₈H₁₆N₂O₄S
Density: 1.24 (20 °C)
Flash Pt: 162 °C
MDL Number: MFCD06798189
CAS Number: 342573-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14adbf116dabf419509d73a392f21ec9883ef10f&vid=14adbf116dabf419509d73a392f21ec9883ef10f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR