2-(5-Ethylpyridin-2-yl)ethanol ≥98.0%

Supplier: TCI America

E0562-25G E0562-5G
TCE0562-25GEA 127.98 CAD
TCE0562-25G TCE0562-5G
2-(5-Ethylpyridin-2-yl)ethanol ≥98.0%
2-(5-Ethylpyridin-2-yl)ethanol

CAS Number: 5223-06-3
MDL Number: MFCD00129040
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 44


Other Product Names / Synonyms: 5-Ethyl-2-(2-hydroxyethyl)pyridine, 2-(5-Ethyl-2-pyridyl)ethanol

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Melting Pt: 44 °C
MDL Number: MFCD00129040
CAS Number: 5223-06-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df9995bcc2c54fdb5b3a6b1c38d8984023f5b28b&vid=df9995bcc2c54fdb5b3a6b1c38d8984023f5b28b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR