1-Ethyl-3-methylimidazolium bromide ≥98.0%

Supplier: TCI America

Synonyms: 3-Ethyl-1-methyl-1H-imidazol-3-ium bromide

E0543-5G E0543-25G
TCE0543-5GEA 52.14 CAD
TCE0543-5G TCE0543-25G
1-Ethyl-3-methylimidazolium bromide ≥98.0%
1-Ethyl-3-methylimidazolium bromide

CAS Number: 65039-08-9
MDL Number: MFCD03427610
Molecular Formula: C6H11BrN2
Molecular Weight: 191.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 74

Formula: C₆H₁₁BrN₂
MW: 191.07 g/mol
Melting Pt: 74 °C
MDL Number: MFCD03427610
CAS Number: 65039-08-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f844f3cf6660d5bdb22d2a654a11c2bbe031507&vid=9f844f3cf6660d5bdb22d2a654a11c2bbe031507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR