7-Ethoxycoumarin ≥97.0%

Supplier: TCI America

E0538-5G E0538-25G
TCE0538-5GEA 232.26 CAD
TCE0538-5G TCE0538-25G
7-Ethoxycoumarin ≥97.0%
7-Ethoxycoumarin

CAS Number: 31005-02-4
MDL Number: MFCD00006877
Molecular Formula: C11H10O3
Molecular Weight: 190.20
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 90

Formula: C₁₁H₁₀O₃
Melting Pt: 90 °C
MDL Number: MFCD00006877
CAS Number: 31005-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37a722e80a0bfc2ec213e1457ef4eb20ee91131e&vid=37a722e80a0bfc2ec213e1457ef4eb20ee91131e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR