N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine ≥98.0%

Supplier: TCI America

Synonyms: N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, NEPA, N-[(S)-1-Carboxyethyl]-L-homophenylalanine Ethyl Ester, (S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoic acid, N-(1S)-Ethoxycarbonyl-3-phenylpropyl-(S)-alanine

E0530-25G E0530-5G
TCE0530-25GEA 107.44 CAD
TCE0530-25G TCE0530-5G
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine ≥98.0%
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine

CAS Number: 82717-96-2
MDL Number: MFCD00209976
Molecular Formula: C15H21NO4
Molecular Weight: 279.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: 29.5 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-[(S)-1-Carboxyethyl]-L-homophenylalanine Ethyl Ester

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00209976
CAS Number: 82717-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cea096c4fa2d835ced4ed2f7001f60abdabc37d4&vid=cea096c4fa2d835ced4ed2f7001f60abdabc37d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR