Ethyl-N-phenylformimidate ≥98.0%

Supplier: TCI America

E0495-5G E0495-25G
TCE0495-005GEA 47.4 CAD
TCE0495-005G TCE0495-025G
Ethyl-N-phenylformimidate ≥98.0%
Ethyl-N-phenylformimidate

CAS Number: 6780-49-0
MDL Number: MFCD00043655
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Specific Gravity (20/20): 1.01


Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: N-Ethoxymethyleneaniline, N-Phenylformimidic Acid Ethyl Ester

Formula: C₉H₁₁NO
Boiling Pt: 215 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00043655
CAS Number: 6780-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15a9c47556d35e3a0cc822f6a1002101d92de4ae&vid=15a9c47556d35e3a0cc822f6a1002101d92de4ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR