3-Ethyl-2-methylbenzoxazolium iodide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

E0477-5G E0477-25G
TCE0477-5GEA 164.32 CAD
TCE0477-5G TCE0477-025G
3-Ethyl-2-methylbenzoxazolium iodide ≥98.0% (by HPLC, titration analysis)
3-Ethyl-2-methylbenzoxazolium iodide

CAS Number: 5260-37-7
MDL Number: MFCD00044141
Molecular Formula: C10H12INO
Molecular Weight: 289.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 197

Formula: C₁₀H₁₂INO
Melting Pt: 197 °C
MDL Number: MFCD00044141
CAS Number: 5260-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b0b02b21698468733c359df7c2681216493f5df&vid=0b0b02b21698468733c359df7c2681216493f5df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR