1,2-Bis(tosyloxy)ethane ≥99.0%

Supplier: TCI America

Synonyms: Ethane-1,2-diyl bis(4-methylbenzenesulphonate), Ethylene glycol bis-p-toluenesulfonate, Ethylene Glycol Ditosylate, 1,2-Bis(p-toluenesulphonyloxy)ethane, Ethane-1,2-diyl bis(4-methylbenzenesulfonate), Ethylene Glycol Bis(p-toluenesulphonate), 1,2-Bis(p-toluenesulfonyloxy)ethane, Ethylene Glycol Bis(p-toluenesulfonate), Ethylene glycol bis-p-toluenesulphonate

E0438-25G E0438-100G
TCE0438-025GEA 123.24 CAD
TCE0438-025G 76220-006
1,2-Bis(tosyloxy)ethane ≥99.0%
1,2-Bis(tosyloxy)ethane

CAS Number: 6315-52-2
MDL Number: MFCD00008549
Molecular Formula: C16H18O6S2
Molecular Weight: 370.43
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 128


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 1,2-Bis(p-toluenesulfonyloxy)ethane, Ethylene Glycol Bis(p-toluenesulfonate), Ethylene Glycol Ditosylate

Formula: C₁₆H₁₈O₆S₂
MW: 370.43 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00008549
CAS Number: 6315-52-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4b103fee96d97bcb10b65fe0f1572e210c6f21a&vid=f4b103fee96d97bcb10b65fe0f1572e210c6f21a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR