exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
E0392-25G
E0392-100G
TCE0392-025GEA
90.06
CAD
TCE0392-025G
TCE0392-500G
TCE0392-100G
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98.0% (by titrimetric analysis)
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride
CAS Number: 6118-51-0
MDL Number: MFCD00151506
Molecular Formula: C8H6O4
Molecular Weight: 166.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Anhydride
Formula:
C₈H₆O₄ |
MDL Number:
MFCD00151506 CAS Number: 6118-51-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...