3-Ethoxy-2-cyclohexen-1-one ≥98.0%
Supplier: TCI America
E0384-5ML
E0384-25ML
TCE0384-5MLEA
77.17
CAD
TCE0384-5ML
TCE0384-025ML
3-Ethoxy-2-cyclohexen-1-one ≥98.0%
3-Ethoxy-2-cyclohexen-1-one
CAS Number: 5323-87-5
MDL Number: MFCD00001580
Molecular Formula: C8H12O2
Molecular Weight: 140.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 107
Specific Gravity (20/20): 1.05
Formula:
C₈H₁₂O₂ Boiling Pt: 119 °C Density: 1.05 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00001580 CAS Number: 5323-87-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...