Ellagic acid dihydrate ≥98.0%
Supplier: TCI America
Synonyms:
2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione
CAS Number: 133039-73-3
MDL Number: MFCD00006914
Molecular Formula: C14H6O8
Molecular Weight: 302.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 450
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: 2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione
Formula:
C₁₄H₆O₈ MW: 302.19 g/mol Melting Pt: 450 °C |
MDL Number:
MFCD00006914 CAS Number: 133039-73-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...