2-Ethynylpyridine ≥97.0%
Supplier: TCI America
E0340-1ML
E0340-5ML
TCE0340-1MLEA
58.46
CAD
TCE0340-1ML
TCE0340-005ML
2-Ethynylpyridine ≥97.0%
2-Ethynylpyridine
CAS Number: 1945-84-2
MDL Number: MFCD00041598
Molecular Formula: C7H5N
Molecular Weight: 103.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 63
Specific Gravity (20/20): 1.02
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 2-Pyridylacetylene
Formula:
C₇H₅N Boiling Pt: 90 °C Density: 1.02 (20 °C) Flash Pt: 63 °C |
MDL Number:
MFCD00041598 CAS Number: 1945-84-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...