8-Ethoxyquinoline-5-sulfonic acid hydrate ≥98.0%

Supplier: TCI America

Synonyms: Actinoquinol

E0339-5G E0339-25G
TCE0339-5GEA 44.24 CAD
TCE0339-5G TCE0339-025G
8-Ethoxyquinoline-5-sulfonic acid hydrate ≥98.0%
8-Ethoxyquinoline-5-sulfonic acid hydrate

CAS Number: 15301-40-3
MDL Number: MFCD00065059
Molecular Formula: C11H11NO4S
Molecular Weight: 253.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: Actinoquinol

Formula: C₁₁H₁₁NO₄S
MW: 253.27 g/mol
MDL Number: MFCD00065059
CAS Number: 15301-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a9f6050664ac1d66167ef9e9df6e45a37fdce2f&vid=4a9f6050664ac1d66167ef9e9df6e45a37fdce2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR