Eicosanedioic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
E0320-250G
E0320-25G
TCE0320-250GEA
1669.43
CAD
TCE0320-250G
TCE0320-025G
Eicosanedioic acid ≥98.0% (by titrimetric analysis)
Eicosanedioic acid
CAS Number: 2424-92-2
MDL Number: MFCD00059627
Molecular Formula: C20H38O4
Molecular Weight: 342.52
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 234
Melting point (°C): 127
Formula:
C₂₀H₃₈O₄ Boiling Pt: 234 °C Melting Pt: 127 °C |
MDL Number:
MFCD00059627 CAS Number: 2424-92-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...