Ethylene glycol mono-m-tolyl ether ≥98.0%

Supplier: TCI America

E0302-25ML E0302-5ML
TCE0302-025MLEA 755.24 CAD
TCE0302-025ML TCE0302-005ML
Ethylene glycol mono-m-tolyl ether ≥98.0%
Ethylene glycol mono-m-tolyl ether

CAS Number: 13605-19-1
MDL Number: MFCD00014035
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 3-(2-Hydroxyethoxy)toluene, 2-(3-Methylphenoxy)ethanol, 2-(m-Tolyloxy)ethanol

Formula: C₉H₁₂O₂
Boiling Pt: 146 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00014035
CAS Number: 13605-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7633baae349c95ebef28829167230d8886912a1&vid=f7633baae349c95ebef28829167230d8886912a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR