Ethyl cellulose, 90-110 mPa.s (25 °C) 5% in toluene/ethanol (80:20)

Supplier: TCI America

Synonyms: Ethylcellulose

E0290-25G E0290-500G
TCE0290-025GEA 57.4 CAD
TCE0290-025G TCE0290-500G
Ethyl cellulose, 90-110 mPa.s (25 °C) 5% in toluene/ethanol (80:20)
Ethyl cellulose

CAS Number: 9004-57-3
MDL Number: MFCD00131037
Form: Crystal
Color: White


Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Cellulose, Natural Polymers (for DDS)

MDL Number: MFCD00131037
CAS Number: 9004-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR