Sodium ethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E0256-25G E0256-5G
TCE0256-025GEA 361.63 CAD
TCE0256-025G TCE0256-005G
Sodium ethanesulfonate ≥98.0% (by titrimetric analysis)
Sodium ethanesulfonate

CAS Number: 5324-47-0
MDL Number: MFCD00066498
Molecular Formula: C2H6O3S
Molecular Weight: 132.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 270


Other Product Names / Synonyms: Ethylsulfonic Acid Sodium Salt, Ethanesulfonic Acid Sodium Salt

Formula: C₂H₆O₃S
Melting Pt: 270 °C
MDL Number: MFCD00066498
CAS Number: 5324-47-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d2c9e4ccd0f0f950be02f6d4ef6bf61d63c2240&vid=4d2c9e4ccd0f0f950be02f6d4ef6bf61d63c2240&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR