Potassium-2-ethylhexanoate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Potassium 2-ethylhexanoate, 2-Ethylhexanoic Acid Potassium Salt
E0121-25G
E0121-500G
TCE0121-025GEA
106.21
CAD
TCE0121-025G
TCE0121-500G
Potassium-2-ethylhexanoate ≥95.0% (by titrimetric analysis)
Potassium-2-ethylhexanoate
CAS Number: 3164-85-0
MDL Number: MFCD00045896
Molecular Formula: C8H16O2
Molecular Weight: 182.30
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2-Ethylhexanoic Acid Potassium Salt
Formula:
C₈H₁₆O₂ MW: 182.30 g/mol |
MDL Number:
MFCD00045896 CAS Number: 3164-85-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...