Ethylenediaminetetraacetic acid disodium zinc salt hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E0098-500G E0098-25G
TCE0098-500GEA 308.1 CAD
TCE0098-500G TCE0098-025G
Ethylenediaminetetraacetic acid disodium zinc salt hydrate ≥98.0% (by titrimetric analysis)
Ethylenediaminetetraacetic acid disodium zinc salt

CAS Number: 14025-21-9
MDL Number: MFCD00135187
Molecular Formula: C10H16N2O8
Molecular Weight: 399.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Disodium Zinc Ethylenediaminetetraacetate

Formula: C₁₀H₁₂N₂Na₂O₈Zn
MDL Number: MFCD00135187
CAS Number: 14025-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fac7b074a3f5f5ce9024fa40e40c61cf9fc88811&vid=fac7b074a3f5f5ce9024fa40e40c61cf9fc88811&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR