Ethyl cellulose, 18-22 mPa.s (25 °C) 5% in toluene/ethanol (80:20)
Supplier: TCI America
E0072-25G
E0072-500G
TCE0072-025GEA
46.63
CAD
TCE0072-025G
TCE0072-500G
Ethyl cellulose, 18-22 mPa.s (25 °C) 5% in toluene/ethanol (80:20)
Ethyl cellulose
CAS Number: 9004-57-3
MDL Number: MFCD00131037
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Cellulose, Natural Polymers (for DDS)
MDL Number:
MFCD00131037 CAS Number: 9004-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...