DL-Ethionine ≥98.0%
Supplier: TCI America
Synonyms:
2-Amino-4-(ethylthio)butanoic acid
E0038-5G
E0038-25G
TCE0038-005GEA
383.94
CAD
TCE0038-005G
TCE0038-025G
DL-Ethionine ≥98.0%
DL-Ethionine
CAS Number: 67-21-0
MDL Number: MFCD00063102
Molecular Formula: C6H13NO2S
Molecular Weight: 163.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₆H₁₃NO₂S MW: 163.24 g/mol |
MDL Number:
MFCD00063102 CAS Number: 67-21-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...