1,2-Ethanedisulfonic acid disodium salt ≥98.0%

Supplier: TCI America

Synonyms: Sodium ethane-1,2-disulfonate, Sodium ethane-1,2-disulphonate

E0031-25G E0031-5G
TCE0031-025GEA 219.62 CAD
TCE0031-025G TCE0031-5G
1,2-Ethanedisulfonic acid disodium salt ≥98.0%
1,2-Ethanedisulphonic acid disodium salt

CAS Number: 5325-43-9
MDL Number: MFCD00024998
Molecular Formula: C2H6O6S2
Molecular Weight: 234.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1,2-Ethanedisulfonic Acid Disodium Salt

Formula: C₂H₄Na₂O₆S₂
MW: 234.1600 g/mol
MDL Number: MFCD00024998
CAS Number: 5325-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dec8f46ef5bc5f5dad34c7dc5dc4ebba09d69c94&vid=dec8f46ef5bc5f5dad34c7dc5dc4ebba09d69c94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR