meso-Erythritol ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: meso-Erythritol, (2R,3S)-rel-Butane-1,2,3,4-tetraol, i-Erythritol

E0021-500G E0021-25G E0021-100G
TCE0021-500GEA 576.92 CAD
TCE0021-500G TCE0021-025G TCE0021-100G
meso-Erythritol ≥99.0% (by HPLC, titration analysis)
meso-Erythritol

CAS Number: 149-32-6
MDL Number: MFCD00004710
Molecular Formula: C4H10O4
Molecular Weight: 122.12
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 331
Melting point (°C): 120

Formula: C₄H₁₀O₄
MW: 122.12 g/mol
Boiling Pt: 331 °C
Melting Pt: 120 °C
MDL Number: MFCD00004710
CAS Number: 149-32-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=730d903927a2c989bbc86e0d859e8e2e362ecd4c&vid=730d903927a2c989bbc86e0d859e8e2e362ecd4c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR