DSS (Disuccinimidyl suberate) ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D3895-5G D3895-1G
TCD3895-5GEA 513.5 CAD
TCD3895-5G TCD3895-1G
DSS (Disuccinimidyl suberate) ≥98.0% (by HPLC, total nitrogen)
DSS (Disuccinimidyl suberate)

CAS Number: 68528-80-3
MDL Number: MFCD00049059
Molecular Formula: C16H20N2O8
Molecular Weight: 368.34
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 171


Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: Suberic Acid Di(N-succinimidyl) Ester, Di(N-succinimidyl) Octanedioate, Octanedioic Acid Di(N-succinimidyl) Ester, DSS

Formula: C₁₆H₂₀N₂O₈
Melting Pt: 171 °C
MDL Number: MFCD00049059
CAS Number: 68528-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c47245812b471bf12a3509a1d99cfc947ffe0e88&vid=c47245812b471bf12a3509a1d99cfc947ffe0e88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR